Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MAP70-2
Entrez Gene ID
839083
Gene Name
microtubule-associated proteins 70-2
Gene Aliases
AT1G24764, ATMAP70-2, F5A9.19, F5A9_19, microtubule-associated proteins 70-2
Chromosome Location
Chr.1: 8759533 - 8763469 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001332663.1 NP_001322233.1 2 - 3 648 86
NM_001332664.1 NP_001322232.1 3 - 4 627 86
NM_102311.7 NP_173873.5 3 - 4 568 86
GenBank mRNA AY128287.1 - 3 - 4 513 86
BT000826.1 - 3 - 4 414 86

View Full Product Details