Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ECT9
Entrez Gene ID
839689
Gene Name
evolutionarily conserved C-terminal region 9
Gene Aliases
AT1G27960, F13K9.7, F13K9_7, evolutionarily conserved C-terminal region 9
Chromosome Location
Chr.1: 9742137 - 9745796 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001332793.1 NP_001322517.1 6 - 7 1807 139
NM_001332794.1 NP_001322518.1 4 - 5 1657 139
NM_102561.5 NP_174117.2 7 - 8 1515 139
GenBank mRNA BT022109.1 - 7 - 8 1409 139

View Full Product Details