Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CAC2
Entrez Gene ID
833497
Gene Name
acetyl Co-enzyme a carboxylase biotin carboxylase subunit
Gene Aliases
ACETYL-COA CARBOXYLASE, AT5G35360, T26D22.8, T26D22_8, acetyl Co-enzyme a carboxylase biotin carboxylase subunit
Chromosome Location
Chr.5: 13584104 - 13588496 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001203493.1 NP_001190422.1 13 - 14 1649 132
NM_122927.4 NP_198386.1 13 - 14 1651 132
GenBank mRNA AY142630.1 - 13 - 14 1455 132
U90879.1 - 13 - 14 1573 132

View Full Product Details