Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
AHP3
Entrez Gene ID
833930
Gene Name
histidine-containing phosphotransmitter 3
Gene Aliases
ARABIDOPSIS THALIANA HISTIDINE-CONTAINING PHOSPHOTRANSMITTER 2, AT5G39340, ATHP2, HIS-ASP PHOSPHOTRANSFER SIGNAL TRANSDUCER, MUL8.20, MUL8_20, histidine-containing phosphotransmitter 3
Chromosome Location
Chr.5: 15748731 - 15750891 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001344287.1 NP_001318703.1 3 - 4 369 127
NM_001344288.1 NP_001330682.1 3 - 4 482 127
NM_001344289.1 NP_001330681.1 3 - 4 473 127
NM_123296.4 NP_198750.2 3 - 4 380 127
GenBank mRNA AB015143.1 - 3 - 4 286 127
AY050944.1 - 3 - 4 320 127
AY091174.1 - 3 - 4 272 127

View Full Product Details