Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DEG13
Entrez Gene ID
834054
Gene Name
protease Do-like protein
Gene Aliases
AT5G40560, DegP protease 13, DegP13, MNF13.8, MNF13_8, degradation of periplasmic proteins 13
Chromosome Location
Chr.5: 16243537 - 16246519 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001344343.1 NP_001318714.1 7 - 8 1168 122

View Full Product Details