Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
POLA3
Entrez Gene ID
834193
Gene Name
DNA primase POLA3
Gene Aliases
AT5G41880, K16L22.17, K16L22_17, POLA4
Chromosome Location
Chr.5: 16760846 - 16764276 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_123553.6 NP_199003.5 11 - 12 1088 110

View Full Product Details