Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- RPN12b
- Entrez Gene ID
-
834209
- Gene Name
- regulatory particle non-ATPase 12B
- Gene Aliases
-
AT5G42040, MJC20.14, MJC20_14, regulatory particle non-ATPase 12B
- Chromosome Location
-
Chr.5: 16813565 - 16814749 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_123569.3 |
NP_199019.2 |
2 - 3 |
447 |
111 |
View Full Product Details