Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MYA2
Entrez Gene ID
834412
Gene Name
myosin 2
Gene Aliases
ARABIDOPSIS MYOSIN 2, AT5G43900, ATMYA2, F6B6.4, F6B6_4, MYOSIN X1 2, MYOSIN XI-6, XI-2, XI-6, myosin 2
Chromosome Location
Chr.5: 17656894 - 17667451 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001203536.1 NP_001190465.1 35 - 36 4246 151
NM_001203537.1 NP_001190466.1 35 - 36 4237 151
NM_123757.5 NP_199203.1 33 - 34 4338 151

View Full Product Details