Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RRS1
Entrez Gene ID
834562
Gene Name
Disease resistance protein (TIR-NBS-LRR class)
Gene Aliases
ARABIDOPSIS THALIANA WRKY DOMAIN PROTEIN 52, AT5G45260, ATWRKY52, K9E15.2, K9E15_2, RESISTANT TO RALSTONIA SOLANACEARUM 1, SENSITIVE TO LOW HUMIDITY 1, SLH1
Chromosome Location
Chr.5: 18326203 - 18332920 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001085246.2 NP_001078715.1 2 - 3 1561 79
NM_001344604.1 NP_001318742.1 2 - 3 1561 79
NM_123894.4 NP_199339.2 2 - 3 1561 79
GenBank mRNA AK226456.1 - 2 - 3 1562 79

View Full Product Details