Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
APG7
Entrez Gene ID
834630
Gene Name
ThiF family protein
Gene Aliases
AT5G45900, ATAPG7, ATATG7, ATG7, AUTOPHAGY 7, AUTOPHAGY-RELATED 7, K15I22.10, K15I22_10, PEROXISOME UNUSUAL POSITIONING 4, PEUP4
Chromosome Location
Chr.5: 18615175 - 18618597 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_123958.3 NP_568652.1 10 - 11 2047 112
GenBank mRNA AB073173.1 - 10 - 11 2000 112
AF492761.1 - 10 - 11 1918 112
AY034945.1 - 10 - 11 1978 112
AY150456.1 - 10 - 11 1918 112

View Full Product Details