Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DELTA-OAT
Entrez Gene ID
834660
Gene Name
ornithine-delta-aminotransferase
Gene Aliases
AT5G46180, MCL19.24, MCL19_24, ornithine-delta-aminotransferase
Chromosome Location
Chr.5: 18718417 - 18721744 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_123987.4 NP_199430.1 5 - 6 1193 88
GenBank mRNA BT023421.1 - 5 - 6 733 88
BT029160.1 - 5 - 6 720 88

View Full Product Details