Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TIP2;3
Entrez Gene ID
834794
Gene Name
tonoplast intrinsic protein 2;3
Gene Aliases
ARABIDOPSIS THALIANA TONOPLAST INTRINSIC PROTEIN 2;3, AT5G47450, ATTIP2;3, DELTA-TIP3, DELTA-TONOPLAST INTRINSIC PROTEIN 3, MNJ7.4, MNJ7_4, tonoplast intrinsic protein 2;3
Chromosome Location
Chr.5: 19248249 - 19249622 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_124117.3 NP_199556.1 1 - 2 535 74
GenBank mRNA BT011212.1 - 1 - 2 407 74
BT011663.1 - 1 - 2 379 74

View Full Product Details