Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ADG1
Entrez Gene ID
834883
Gene Name
ADP glucose pyrophosphorylase 1
Gene Aliases
ADP GLUCOSE PYROPHOSPHORYLASE SMALL SUBUNIT, ADP glucose pyrophosphorylase 1, ADP glucose pyrophosphorylase 1, ADP-GLUCOSE PYROPHOSPHORYLASE SMALL SUBUNIT 1, APS1, AT5G48300, K23F3.2, K23F3_2
Chromosome Location
Chr.5: 19570238 - 19572804 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_124205.4 NP_199641.1 5 - 6 1316 60
GenBank mRNA AY049265.1 - 5 - 6 1292 60
AY065428.1 - 5 - 6 1258 60
AY090283.1 - 5 - 6 1228 60
AY096379.1 - 5 - 6 1228 60
U72351.1 - 5 - 6 1263 60

View Full Product Details