Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
XRI1
Entrez Gene ID
834930
Gene Name
x-ray induced transcript 1
Gene Aliases
AT5G48720, K24G6.5, K24G6_5, X-RAY INDUCED TRANSCRIPT, X-RAY INDUCED TRANSCRIPT 1, XRI
Chromosome Location
Chr.5: 19758881 - 19762015 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001161298.2 NP_001154770.1 7 - 8 734 94
NM_001344803.1 NP_001330159.1 6 - 7 675 94
NM_001344804.1 NP_001330160.1 6 - 7 622 94
NM_124249.3 NP_199683.2 6 - 7 907 94
GenBank mRNA BT012214.1 - 6 - 7 571 94
BT020567.1 - 4 - 5 406 94

View Full Product Details