Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- PANC
- Entrez Gene ID
-
834942
- Gene Name
- pantoate-beta-alanine ligase
- Gene Aliases
-
ARABIDOPSIS THALIANA PANTOTHENATE SYNTHETASE, AT5G48840, ATPTS, K24G6.18, K24G6_18, PANTOTHENATE SYNTHETASE, PS, PTS, homolog of bacterial PANC
- Chromosome Location
-
Chr.5: 19803502 - 19805356 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_124261.4 |
NP_199695.1 |
2 - 3 |
993 |
75 |
GenBank mRNA |
AK175238.1 |
- |
2 - 3 |
706 |
75 |
View Full Product Details