Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PNG1
Entrez Gene ID
835019
Gene Name
peptide-N-glycanase 1
Gene Aliases
AT5G49570, AtPNG1, K6M13.12, K6M13_12, peptide-N-glycanase 1
Chromosome Location
Chr.5: 20111859 - 20116269 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_124335.3 NP_199768.1 16 - 17 2223 94
GenBank mRNA AK228156.1 - 16 - 17 2143 94
AM745095.1 - 16 - 17 2079 94
AY140065.1 - 16 - 17 2139 94
BT003398.1 - 16 - 17 2079 94

View Full Product Details