Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HSD6
Entrez Gene ID
835149
Gene Name
hydroxysteroid dehydrogenase 6
Gene Aliases
AT5G50770, AtHSD6, MFB16.17, MFB16_17, hydroxysteroid dehydrogenase 6
Chromosome Location
Chr.5: 20646680 - 20648725 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_124455.2 NP_199890.1 5 - 6 788 87
GenBank mRNA BT020525.1 - 5 - 6 780 87

View Full Product Details