Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CRL
Entrez Gene ID
835175
Gene Name
crumpled leaf
Gene Aliases
AT5G51020, CAA33, CRUMPLED LEAF, K3K7.20, K3K7_20, constitutive activator of AAA-ATPase
Chromosome Location
Chr.5: 20745349 - 20747305 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_124481.4 NP_199915.1 7 - 8 712 102
GenBank mRNA AB120415.1 - 7 - 8 649 102
AF370196.1 - 7 - 8 652 102
AY113932.1 - 7 - 8 572 102

View Full Product Details