Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PABN1
Entrez Gene ID
835186
Gene Name
polyadenylate-binding protein 1
Gene Aliases
AT5G51120, ATPABN1, MWD22.6, MWD22_6, POLYADENYLATE-BINDING PROTEIN 1, polyadenylate-binding protein 1
Chromosome Location
Chr.5: 20779327 - 20781557 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001203582.1 NP_001190511.1 6 - 7 853 69
NM_001203583.1 NP_001190512.1 4 - 5 404 69
NM_124491.3 NP_568751.1 5 - 6 858 69
GenBank mRNA AY054186.1 - 5 - 6 699 69
AY057580.1 - 5 - 6 686 69
AY079392.1 - 5 - 6 640 69
AY116673.1 - 5 - 6 683 69

View Full Product Details