Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MSBP1
Entrez Gene ID
835300
Gene Name
membrane steroid binding protein 1
Gene Aliases
ARABIDOPSIS THALIANA MEMBRANE-ASSOCIATED PROGESTERONE BINDING PROTEIN 5, AT5G52240, ATMP1, AtMAPR5, F17P19.14, F17P19_14, membrane steroid binding protein 1
Chromosome Location
Chr.5: 21212940 - 21214842 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001203592.2 NP_001190521.1 1 - 2 463 95
NM_124603.4 NP_200037.1 1 - 2 503 95
GenBank mRNA BT000922.1 - 1 - 2 322 95

View Full Product Details