Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MLO11
Entrez Gene ID
835457
Gene Name
Seven transmembrane MLO family protein
Gene Aliases
AT5G53760, ATMLO11, MGN6.12, MGN6_12, MILDEW RESISTANCE LOCUS O 11
Chromosome Location
Chr.5: 21822323 - 21826488 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001036993.2 NP_001032070.1 3 - 4 695 108
NM_124755.3 NP_200187.1 2 - 3 785 108
GenBank mRNA AF369572.1 - 1 - 2 144 108
AK317279.1 - 3 - 4 485 108
AY057502.1 - 2 - 3 629 108
BT000434.1 - 2 - 3 619 108
BT002581.1 - 1 - 2 144 108

View Full Product Details