Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- ILL3
- Entrez Gene ID
-
835501
- Gene Name
- IAA-leucine-resistant (ILR1)-like 3
- Gene Aliases
-
AT5G54140, IAA-leucine-resistant (ILR1)-like 3, MJP23.12, MJP23_12
- Chromosome Location
-
Chr.5: 21965667 - 21968061 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_124794.3 |
NP_200225.1 |
1 - 2 |
488 |
125 |
View Full Product Details