Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SIGF
Entrez Gene ID
818273
Gene Name
RNApolymerase sigma-subunit F
Gene Aliases
AT2G36990, ATSIG6, RNApolymerase sigma-subunit F, SIG6, SIGMA FACTOR 6, SOLDAT8, T1J8.17, T1J8_17
Chromosome Location
Chr.2: 15537225 - 15540139 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_129255.3 NP_565856.1 4 - 5 643 98
GenBank mRNA AB029916.1 - 4 - 5 520 98
AJ277389.1 - 4 - 5 602 98
AY059768.1 - 4 - 5 643 98
AY091182.1 - 4 - 5 520 98

View Full Product Details