Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
VCL1
Entrez Gene ID
818380
Gene Name
vacuoleless1 (VCL1)
Gene Aliases
AT2G38020, EMB258, MAN, MANGLED, T8P21.7, T8P21_7, VACUOLELESS 1
Chromosome Location
Chr.2: 15909908 - 15914711 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001084551.1 NP_001078020.1 12 - 13 2196 101
NM_129359.4 NP_565879.1 12 - 13 2197 101
GenBank mRNA AF359240.1 - 11 - 12 2104 101
AY048248.1 - 12 - 13 2171 101
AY139799.1 - 11 - 12 2088 101

View Full Product Details