Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MAP65-5
Entrez Gene ID
818454
Gene Name
microtubule-associated protein 65-5
Gene Aliases
AT2G38720, T6A23.8, T6A23_8, microtubule-associated protein 65-5
Chromosome Location
Chr.2: 16188047 - 16192102 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001336724.1 NP_001318376.1 5 - 6 867 113

View Full Product Details