Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MLO12
Entrez Gene ID
818505
Gene Name
Seven transmembrane MLO family protein
Gene Aliases
AT2G39200, ATMLO12, MILDEW RESISTANCE LOCUS O 12, T16B24.16, T16B24_16
Chromosome Location
Chr.2: 16356041 - 16359943 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_129478.5 NP_565902.1 7 - 8 903 96
GenBank mRNA AF369573.1 - 7 - 8 773 96

View Full Product Details