Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TOPP4
Entrez Gene ID
818571
Gene Name
type one serine/threonine protein phosphatase 4
Gene Aliases
AT2G39840, type one serine/threonine protein phosphatase 4
Chromosome Location
Chr.2: 16627216 - 16629412 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_129543.3 NP_181514.1 2 - 3 997 130
GenBank mRNA AK229028.1 - 2 - 3 824 130
BT026118.1 - 2 - 3 783 130
M93411.1 - 2 - 3 804 130

View Full Product Details