Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CSY3
Entrez Gene ID
818879
Gene Name
citrate synthase 3
Gene Aliases
AT2G42790, F7D19.21, F7D19_21, citrate synthase 3
Chromosome Location
Chr.2: 17802841 - 17806073 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_129840.5 NP_181807.1 11 - 12 1345 107
GenBank mRNA AK227008.1 - 11 - 12 1322 107
AY048214.1 - 11 - 12 1321 107
AY124841.1 - 11 - 12 1263 107

View Full Product Details