Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- ETFQO
- Entrez Gene ID
-
818941
- Gene Name
- electron-transfer flavoprotein:ubiquinone oxidoreductase
- Gene Aliases
-
AT2G43400, T1O24.14, electron-transfer flavoprotein:ubiquinone oxidoreductase
- Chromosome Location
-
Chr.2: 18021109 - 18025292 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_129901.4 |
NP_181868.1 |
16 - 17 |
1945 |
64 |
View Full Product Details