Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MLO15
Entrez Gene ID
819017
Gene Name
Seven transmembrane MLO family protein
Gene Aliases
AT2G44110, ATMLO15, F6E13.24, MILDEW RESISTANCE LOCUS O 15
Chromosome Location
Chr.2: 18245357 - 18248459 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_129974.1 NP_181939.1 1 - 2 116 120
NM_201957.2 NP_973686.1 1 - 2 259 120
GenBank mRNA AF369576.1 - 1 - 2 116 120

View Full Product Details