Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TPD1
Entrez Gene ID
2745725
Gene Name
tapetum determinant 1
Gene Aliases
AT4G24972, TAPETUM DETERMINANT 1
Chromosome Location
Chr.4: 12837353 - 12840230 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_202883.3 NP_974612.1 2 - 3 285 74
GenBank mRNA AY394846.1 - 2 - 3 285 74

View Full Product Details