Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
AGP9
Entrez Gene ID
815978
Gene Name
arabinogalactan protein 9
Gene Aliases
ARABINOGALACTAN PROTEIN 9, AT2G14890, T26I20.5, T26I20_5, arabinogalactan protein 9
Chromosome Location
Chr.2: 6399405 - 6401149 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_127053.5 NP_179095.1 1 - 2 780 103
GenBank mRNA AF195890.1 - 1 - 2 549 103
AF326868.1 - 1 - 2 557 103
AF339690.1 - 1 - 2 506 103
AY078968.1 - 1 - 2 564 103
AY125554.1 - 1 - 2 506 103

View Full Product Details