Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- CYP79F2
- Entrez Gene ID
-
838210
- Gene Name
- cytochrome P450, family 79, subfamily F, polypeptide 2
- Gene Aliases
-
''cytochrome P450, AT1G16400, CYTOCHROME P450 79F2, F3O9.20, F3O9_20, cytochrome P450, family 79, polypeptide 2, polypeptide 2'', subfamily F
- Chromosome Location
-
Chr.1: 5605044 - 5607498 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_101506.3 |
NP_563995.2 |
2 - 3 |
1433 |
111 |
GenBank mRNA |
AF370512.1 |
- |
2 - 3 |
1320 |
111 |
View Full Product Details