Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PP2A-1
Entrez Gene ID
842276
Gene Name
protein phosphatase 2A-2
Gene Aliases
AT1G59830, F23H11.15, F23H11_15, protein phosphatase 2A-1, protein phosphatase 2A-2
Chromosome Location
Chr.1: 22020377 - 22022503 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_104676.5 NP_176192.1 2 - 3 571 117
NM_202321.2 NP_974050.1 2 - 3 552 117
GenBank mRNA AY063942.1 - 2 - 3 513 117
AY096543.1 - 2 - 3 361 117
M96732.1 - 2 - 3 451 117

View Full Product Details