Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MLP43
Entrez Gene ID
843427
Gene Name
MLP-like protein 43
Gene Aliases
AT1G70890, F15H11.12, F15H11_12, MLP-like protein 43
Chromosome Location
Chr.1: 26725521 - 26726652 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_105756.4 NP_177245.1 1 - 2 355 131
GenBank mRNA AY065135.1 - 1 - 2 247 131
BT008441.1 - 1 - 2 192 131

View Full Product Details