Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- NAP5
- Entrez Gene ID
-
843474
- Gene Name
- non-intrinsic ABC protein 5
- Gene Aliases
-
AT1G71330, ATNAP5, F3I17.2, F3I17_2, non-intrinsic ABC protein 5
- Chromosome Location
-
Chr.1: 26884014 - 26885169 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_105802.1 |
NP_177289.1 |
1 - 2 |
225 |
150 |
View Full Product Details