Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- PME7
- Entrez Gene ID
-
841063
- Gene Name
- pectin methylesterase 7
- Gene Aliases
-
A. THALIANA PECTIN METHYLESTERASE 7, AT1G44980, ATPME7, F27F5.7, F27F5_7, pectin methylesterase 7
- Chromosome Location
-
Chr.1: 17004256 - 17005923 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_103578.2 |
NP_175118.2 |
3 - 4 |
476 |
146 |
View Full Product Details