Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GA20OX4
Entrez Gene ID
842389
Gene Name
gibberellin 20-oxidase 4
Gene Aliases
AT1G60980, ATGA20OX4, T7P1.12, T7P1_12, gibberellin 20-oxidase 4
Chromosome Location
Chr.1: 22452573 - 22454140 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_104778.1 NP_176294.1 2 - 3 876 159
GenBank mRNA DQ056503.1 - 2 - 3 876 159

View Full Product Details