Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RPS15AD
Entrez Gene ID
823747
Gene Name
ribosomal protein S15A D
Gene Aliases
AT3G46040, ribosomal protein S15A D
Chromosome Location
Chr.3: 16914525 - 16915837 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_114473.6 NP_190190.1 3 - 4 428 162
GenBank mRNA AY045837.1 - 3 - 4 387 162
AY091373.1 - 2 - 3 294 162

View Full Product Details