Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
scpl36
Entrez Gene ID
824363
Gene Name
serine carboxypeptidase-like 36
Gene Aliases
AT3G52000, serine carboxypeptidase-like 36
Chromosome Location
Chr.3: 19293010 - 19294881 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_115059.3 NP_190768.1 2 - 3 512 115
GenBank mRNA DQ056620.1 - 2 - 3 451 115

View Full Product Details