Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
WRKY69
Entrez Gene ID
825040
Gene Name
WRKY DNA-binding protein 69
Gene Aliases
AT3G58710, ATWRKY69, RABIDOPSIS THALIANA WRKY DNA-BINDING PROTEIN 69, WRKY DNA-binding protein 69
Chromosome Location
Chr.3: 21714911 - 21716916 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_115733.5 NP_567073.2 1 - 2 406 116
GenBank mRNA AF421156.1 - 1 - 2 125 116

View Full Product Details