Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RGXT2
Entrez Gene ID
827905
Gene Name
rhamnogalacturonan xylosyltransferase 2
Gene Aliases
AT4G01750, T7B11.1, rhamnogalacturonan xylosyltransferase 2
Chromosome Location
Chr.4: 756348 - 758725 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_116405.6 NP_192084.1 2 - 3 854 82
GenBank mRNA AY065071.1 - 2 - 3 840 82
BT002558.1 - 2 - 3 668 82
KJ138772.1 - 2 - 3 668 82

View Full Product Details