Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RAC6
Entrez Gene ID
829750
Gene Name
RAC-like 6
Gene Aliases
ARAC6, AT4G35950, ATRAC6, ATROP5, RAC-like 2, RAC-like 6, RAC2, RHO-RELATED PROTEIN FROM PLANTS 5, ROP5, T19K4.80
Chromosome Location
Chr.4: 17023837 - 17025866 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_119762.4 NP_195320.1 3 - 4 651 93
GenBank mRNA AF079487.1 - 3 - 4 531 93
AF107663.1 - 3 - 4 358 93
AK117209.1 - 3 - 4 648 93
BT005217.1 - 3 - 4 299 93

View Full Product Details