Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
scpl1
Entrez Gene ID
833615
Gene Name
serine carboxypeptidase-like 1
Gene Aliases
AT5G36180, MAB16.13, MAB16_13, serine carboxypeptidase-like 1
Chromosome Location
Chr.5: 14238935 - 14242135 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001344130.1 NP_001331683.1 10 - 11 1536 178
NM_001344132.1 NP_001331685.1 10 - 11 1081 178
NM_123009.4 NP_198467.2 11 - 12 1462 178
GenBank mRNA AY093044.1 - 11 - 12 1220 178
AY128925.1 - 11 - 12 1199 178

View Full Product Details