Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- GSTT3
- Entrez Gene ID
-
834124
- Gene Name
- glutathione S-transferase THETA 3
- Gene Aliases
-
AT5G41220, ATGSTT3, GST10C, K1O13.1, K1O13_1, glutathione S-transferase THETA 3
- Chromosome Location
-
Chr.5: 16494325 - 16497163 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_123487.4 |
NP_198938.1 |
2 - 3 |
337 |
133 |
View Full Product Details