Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
COR15B
Entrez Gene ID
818853
Gene Name
cold regulated 15b
Gene Aliases
AT2G42530, F14N22.20, F14N22_20, cold regulated 15b
Chromosome Location
Chr.2: 17708593 - 17710433 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_129814.3 NP_181781.1 1 - 2 753 168
GenBank mRNA AY045809.1 - 1 - 2 265 168
AY085876.1 - 1 - 2 265 168
AY091440.1 - 1 - 2 193 168
FJ156734.1 - 1 - 2 265 168

View Full Product Details