Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GH9B11
Entrez Gene ID
819064
Gene Name
glycosyl hydrolase 9B11
Gene Aliases
AT2G44560, AtGH9B11, F16B22.5, glycosyl hydrolase 9B11
Chromosome Location
Chr.2: 18391753 - 18393729 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_130020.3 NP_181984.1 3 - 4 1229 98
GenBank mRNA AK175889.1 - 3 - 4 1207 98

View Full Product Details