Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ATU2AF35A
Entrez Gene ID
839657
Gene Name
U2 snRNP auxiliary factor small subunit
Gene Aliases
AT1G27650, T22C5.30
Chromosome Location
Chr.1: 9614551 - 9616574 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001084140.1 NP_001077609.1 2 - 2 966 97
NM_102530.3 NP_174086.1 2 - 2 1178 97
GenBank mRNA AY065202.1 - 2 - 2 1126 97
AY088540.1 - 2 - 2 1154 97
AY090343.1 - 2 - 2 1145 97

View Full Product Details