Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ELF4-L3
Entrez Gene ID
815182
Gene Name
ELF4-like 3
Gene Aliases
AT2G06255, ELF4-like 3
Chromosome Location
Chr.2: 2457573 - 2459940 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_126634.3 NP_565334.1 2 - 2 616 116
GenBank mRNA AY086681.1 - 2 - 2 616 116

View Full Product Details