Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- EXPA25
- Entrez Gene ID
-
833926
- Gene Name
- expansin A25
- Gene Aliases
-
AT5G39300, ATEXP25, ATEXPA25, ATHEXP ALPHA 1.18, EXP25, EXPANSIN 25, K3K3.150, K3K3_150, expansin A25
- Chromosome Location
-
Chr.5: 15737366 - 15738479 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_123292.2 |
NP_198746.2 |
2 - 2 |
428 |
83 |
View Full Product Details